Skip to Content
Merck
All Photos(2)

Key Documents

MABF853

Sigma-Aldrich

Anti-FcgRIV (CD16-2) Antibody, clone 9E9

clone 9E9, from hamster(Armenian)

Synonym(s):

CD16-2, Fcgr4, FcgRIV, Fc gammaRIV, FcgammaRIV, Fcgr3a, CD16-2, Transmembrane receptor CD16-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

hamster (Armenian)

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

9E9, monoclonal

species reactivity

mouse

technique(s)

flow cytometry: suitable
neutralization: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Fcgr4(246256)

General description

Transmembrane receptor CD16-2 (UniProt: Q8R477; also known as CD16-2, Fcgr4, FcgRIV, Fc gammaRIV, FcgammaRIV, Fcgr3a, CD16-2, Transmembrane receptor CD16-2) is encoded by the Fcgr4 (also known as Fcgr3a, Fcrl3) gene (Gene ID: 245256) in murine species. Murine species express three classes of activating IgG Fc receptors, the high affinity receptor FcgRI, the low affinity receptor FcgRIII, and the FcgRIV or CD16-2. CD16-2, a novel mouse receptor that displays significant homology to CD16/FcgRIII. It is a receptor for soluble IgG2a and 2b complexes, but not necessarily for cellular bound IgG2a. CD16-2 is expressed in peripheral blood leukocytes and in spleen, thymus, colon, and intestine. Similar to other activating receptors, CD16-2 possesses a charged residue within its transmembrane domain that is necessary for association with an ITAM (immunoreceptor tyrosine-based activation motif) bearing accessory protein. Both CD16-2 (FcgRIV) and FcgRIII are considered to be essential for mediating type II and type III autoimmune responses via FcRc-LAT-dependent generation of C5a. However, FcgRIII-deficient mice display compensatory enhanced CD16-2 expression that offers protection from lung inflammation and exhibit reduced sensitivity to IgG2a/b-mediated hemolytic anemia. Ref.: Syed, S.N., et al (2009). Eur. J. Immunol. 39(12): 3343-3356; Mechetina, L.V., et al. (2002). Immunogenetics 54(7): 463-468.

The previously assigned protein identifier Q8R477 has been merged into A0A0B4J1G0. Full details can be found on the UniProt database.

Specificity

Clone 9E9 immunostained murine macrophages, neutrophils, dendritic cells, but not FcgRIII-expressing NK cells. Clone 9E9 also stained a FcgRIV-negative, B220-positive peripheral blood cell population (Syed, S.N., et al. (2009). Eur. J. Immunol. 39(12):3343-3356).

Immunogen

Recombinant mouse FcgRIV extracellular domain fused to mouse IgG1 Fc fragment (Nimmerjahn, F., et al. (2005). Immunity. 23(1):41-51).

Application

Flow Cytometry Analysis: 20 µg/mL from a representative lot, when added prior to APC-conjugated 9E9, blocked the staining of FcgRIV-positive granulocytes and monocytes by APC-conjugated 9E9 (Courtesy of Jeanette Leusen, Ph.D, University Medical Center, Utrecht, Netherlands).

Flow Cytometry Analysis: A representative lot detected FcgRIV-positive granulocytes and monocytes from wild-type, but not Fcgr4-knockout mice (Nimmerjahn, F., et al. (2010). Proc. Natl. Acad. Sci. U. S. A. 107(45):19396-19401).

Flow Cytometry Analysis: A representative lot, pre-conjugated with Alexa Fluor 647, immunostained murine macrophages, neutrophils, dendritic cells, but not FcgRIII-expressing NK cells. Clone 9E9 also stained a FcgRIV-negative, B220-positive peripheral blood cell population (Syed, S.N., et al. (2009). Eur. J. Immunol. 39(12):3343-3356).

Neutralizing Analysis: A representative lot, when administered (400-500 µg) via i.v. injection prior to anti-RBC 34-3C-IgG2b (50 µg) i.p. injection, prevented anemia (RBC depletion) induction by 34-3C-IgG2b. Clone 9E9 also prevented immunocomplex (IC) challenge-induced alveolar RBC & PMN infiltration in IC-sensitized mice (Syed, S.N., et al. (2009). Eur. J. Immunol. 39(12):3343-3356).

Neutralizing Analysis: A representative lot suppressed both wild-type and Fcgr1-/Fcgr3-knockout mouse peritoneal macrophages-mediated phogocytosis of red blood cells (RBCs) opsonized with anti-RBC IgG2b, but not anti-RBC IgG2a (Syed, S.N., et al. (2009). Eur. J. Immunol. 39(12):3343-3356).

Neutralizing Analysis: A representative lot prevented immune complex binding-induced activation of Fcgr3-deficient mouse neutrophils (Jakus, Z., et al. (2008). J. Immunol. 180(1):618-629).

Neutralizing Analysis: A representative lot, when administered (200 µg) via i.v. injection 30 min prior to anti-platelet 6A6-IgG2a Fc fusion (3.5 µg) injection, reduced the extend of thrombocytopenia (platelet depletion) induction by 6A6-IgG2a (Nimmerjahn, F., et al. (2005). Immunity. 23(1):41-51).
This armenian hamster monoclonal Anti-FcgRIV (CD16-2) Antibody, clone 9E9, Cat. No. MABF853 is validated for use in Flow Cytometry and Neutralization.

Quality

Evaluated by Flow Cytometry in RAW264.7 cells.

Flow Cytometry Analysis: 0.1 µg of this antibody detected FcgRIV on the surface of one million RAW264.7 murine macrophages.

Target description

28.39/28.37/28.38 kDa (UniProt Q3TC44/Q8R2R4/Q8R477) calculated.

Physical form

Format: Purified
Purified armenian hamster IgG in PBS without azide.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service