MABF2155
Anti-Ricin A Chain Antibody, clone RAC18
clone RAC18, from mouse
Synonym(s):
RAC
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
mouse
antibody form
purified antibody
antibody product type
primary antibodies
clone
RAC18, monoclonal
species reactivity
Ricinus communis
packaging
antibody small pack of 25 μg
technique(s)
ELISA: suitable
inhibition assay: suitable
neutralization: suitable
isotype
IgG2aκ
UniProt accession no.
target post-translational modification
unmodified
Related Categories
General description
Ricin is a dimeric toxin derived from Ricinus communis (Caster bean). It is composed of a sugar-binding subunit (B chain) that attached to cell surface receptors and facilitate the entry of toxin into the target cell and a second subunit (A chain), whcih has glycosidase activity that inactivates ribosomes by depurinating a single adenosine residue from an exposed loop of the 28S ribosomal RNA. This leads to inhibition of protein synthesis in cells. The toxic effects of Ricin are attributed to the A chain. Clone RAC18 displays high avidity for intact ricin and inhibits its activity without affecting binding of ricin to cells. Immunization with RAC18 is shown to offer significant protection against ricin toxicity. (Ref.: Maddaloni, M., et al. (2004). J. Immunol. 172; 6221-6228).
Specificity
Clone RAC18 bind to Ricin A chain
Immunogen
Purified Ricin A chain.
Application
Anti-Ricin A Chain, clone RAC18, Cat. No. MABF2155, is a mouse monoclonal antibody that detects Ricin A chain and has been tested for use in ELISA, Inhibition Assay, and Neutralizing.
Neutralizing Analysis: A representative lot neutralized the activity of Ricin A chain. (Maddaloni, M., et. al. (2004). J Immunol. 172(10):6221-8).
ELISA Analysis: A representative lot detected Ricin A chain in ELISA applications (Maddaloni, M., et. al. (2004). J Immunol. 172(10):6221-8; Simon, S., et. al. (2015). Toxins (Basel) 7(12):4967-86).
Inhibition Analysis: A representative lot inhibited the enzymatic activity of Ricin. (Maddaloni, M., et. al. (2004). J Immunol. 172(10):6221-8).
ELISA Analysis: A representative lot detected Ricin A chain in ELISA applications (Maddaloni, M., et. al. (2004). J Immunol. 172(10):6221-8; Simon, S., et. al. (2015). Toxins (Basel) 7(12):4967-86).
Inhibition Analysis: A representative lot inhibited the enzymatic activity of Ricin. (Maddaloni, M., et. al. (2004). J Immunol. 172(10):6221-8).
Research Category
Inflammation & Immunology
Inflammation & Immunology
Quality
Evaluated by ELISA with Ricin A chain.
ELISA Analysis: Various dilutions of this antibody detected Ricin A chain in an indirect ELISA.
ELISA Analysis: Various dilutions of this antibody detected Ricin A chain in an indirect ELISA.
Target description
29.89 kDa calculated.
Physical form
Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG2a in PBS without preservatives.
Storage and Stability
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Other Notes
Concentration: Please refer to lot specific datasheet.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service