Skip to Content
Merck
All Photos(1)

Key Documents

MABF2115

Sigma-Aldrich

Anti-NKR-P1B (CD161b) Antibody, clone 2D12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

antibody form

purified antibody

clone

2D12
monoclonal

purified by

using protein G

species reactivity

mouse, mouse, mouse

concentration

(Please refer to lot specific datasheet.)

technique(s)

flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aλ

suitability

suitable for immunoassay

UniProt accession no.

General description

Mouse BaF/3 cells (murine IL-3-dependent pro-B cell line) expressing mouse NKR-P1B protein.

Specificity

Clone 2D12 is a mouse monoclonal antibody that detects Natural killer cell surface protein NKR-P1B allele B6 (NKR-P1B) in murine species. It does not react with Nkrp1a, Nkrp1c, and Nkrp1f.

Application

Affects Function Analysis: A representative lot reversed the reduction in killing induced by the expression of Clrb in C1498, BW5147, and B6 LAK cells. (Iizuka, K., et. al. (2003). Nat Immunol. 4(8):801-7).

Immunoprecipitation Analysis: A representative lot immunoprecipitated NKR-P1B (CD161b) in Immunoprecipitation applications (Iizuka, K., et. al. (2003). Nat Immunol. 4(8):801-7).

Flow Cytometry Analysis: A representative lot detected NKR-P1B (CD161b) in Flow Cytometry applications (Iizuka, K., et. al. (2003). Nat Immunol. 4(8):801-7; Aguilar, O.A., et. al. (2017). Cell. 169(1):58-71).

Quality

Evaluated by Western Blotting in mouse spleen tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected NKR-P1B (CD161b) in mouse spleen tissue lysate.

Physical properties

~52 kDa observed; 25.03 kDa calculated. . Uncharacterized bands may be observed in some lysate(s).

Physical form

Purified mouse monoclonal antibody IgG2a in PBS without azide.

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service