Skip to Content
Merck
All Photos(1)

Key Documents

MABC588

Sigma-Aldrich

Anti-MgcRacGAP/CYK4 Antibody, clone 5G5

clone 5G5, from mouse

Synonym(s):

Rac GTPase-activating protein 1, Male germ cell RacGap, MgcRacGAP, Protein CYK4 homolog, CYK4, HsCYK-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5G5, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RACGAP1(29127)

General description

MgcRacGAP/CYK4 is a cytoskeletal interacting protein that participates in cellular functions from cell division and differentiation to ion transport, spermatogenesis and protein transport. MgcRacGAP/CYK4 is a component of the multi-protein complex called the centralspindlin complex that is key to cytokinesis function. MgcRacGAP/CYK4 developmentally is key for embryogenesis, adipocyte and myoblast development. It is also key in male germ development. MgcRacGAP/CYK4 associates with various proteins including alpha, beta and gamma tubulin, RACGAP1, and KIF23. MgcRacGAP/CYK4 can be localized in the nucleus but also in the cytoplasm where it is associated with the cytoskeletal spindle and microtubules. MgcRacGAP/CYK4 is highly expressed in testis, thymus and placenta but also expressed in spleen and various lymphocytes and sperm.

Immunogen

Linear peptide corresponding to Human MgcRacGAP/CYK4.

Application

Immunocytochemistry Analysis: A representative lot of this antibody detected MgcRacGAP/CYK4 in HeLa cells (Uehara, R., et al. (2013) JCB. 202(4):623-636).

Immunoprecipitation Analysis: A representative lot of this antibody detected MgcRacGAP/CYK4 by immunoprecipitation.
Research Category
Apoptosis & Cancer
Research Sub Category
Tumor Markers
This Anti-MgcRacGAP/CYK4 Antibody, clone 5G5 is validated for use in Western Blotting and Immunocytochemistry and Immunoprecipitation for the detection of MgcRacGAP/CYK4.

Quality

Evaluated by Western Blotting in HeLa nuclear extract cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected MgcRacGAP/CYK4 in 10 µg of HeLa nuclear extract cell lysate.

Target description

~75 kDa observed

Physical form

Format: Purified
Protein A purified
Purified mouse monoclonal IgG2bκ in buffer containing PBS without preservatives with 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service