Recommended Products
antibody form
purified antibody
clone
376.96
monoclonal
purified by
using protein G
species reactivity
human, human, mouse
concentration
(Please refer to lot specific datasheet.)
technique(s)
flow cytometry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG2aκ
UniProt accession no.
Mode of action
| inhibits
General description
Specificity
Application
Quality
Flow Cytometry Analysis: 1 µg of this antibody detected B7-H3 (CD276) in one million SK-OV-3 cells.
Physical properties
Physical form
Storage and Stability
Disclaimer
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service