Skip to Content
Merck
All Photos(1)

Documents

MAB8674

Sigma-Aldrich

Anti-Herpes Simplex Virus 1 Antibody, clone 13D11

clone 13D11, Chemicon®, from mouse

Synonym(s):

HSV1, HSV, UL42

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

13D11, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

shipped in

wet ice

General description

UL42 is the DNA polymerase accessory protein of herpes simplex virus type 1.

Specificity

Clone 13D11 is specific for the UL42 protein of HSV1

Immunogen

Baculovirus-expressed HSV DNA polymerase (POL) and POL/UL42 complex.

Application

Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral
This Anti-Herpes Simplex Virus 1 Antibody, clone 13D11 is validated for use in ELISA, WB, IH for the detection of Herpes Simplex Virus 1.
Western Blot: predicted molecular weight: 59 kDa on reducing conditions.

Immunohistochemistry: acetone fixed tissues or cell cultures, other fixations not tested.

ELISA

Optimal dilutions must be determined by end user

Linkage

Replaces: MAB8684

Physical form

Format: Purified
Purified immunoglobulin. Liquid 0.02M PB, 0.25M NaCl, pH 7.6 buffer containing 0.1% sodium azide as a preservative.

Storage and Stability

Store at 2 to 8°C for up to 12 months from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service