Skip to Content
Merck
All Photos(1)

Key Documents

MAB5504A4

Sigma-Aldrich

Anti-Vesicular Glutamate Transporter 2 Antibody, Alexa Fluor 488 | MAB5504A4

clone 8G9.2, from mouse, ALEXA FLUOR 488

Synonym(s):

Vesicular glutamate transporter 2, VGluT2, Differentiation-associated BNPI, Differentiation-associated Na(+)-dependent inorganic phosphate cotransporter, Solute carrier family 17 member 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

8G9.2, monoclonal

species reactivity

rat, mouse

species reactivity (predicted by homology)

human (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

General description

Vesicular Glutamate Transporter 2 (VGLUT2) is a sodium dependent inorganic phosphate cotransporter that is involved in the calcium dependent glutamate release from astrocytes. It is expressed in only a subset of neurons, localized to synaptic vesicles, at synapses exhibiting classical excitatory features. VGLUT2 mRNA is found in brain regions that lack VGLUT1, a related protein also displaying sodium dependent inorganic phosphate cotransporter activity.

Immunogen

Recombinant protein from rat VGLUT2.

Application

Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Immunohistochemistry in adult mouse brain tissue.

Immunohistochemistry Analysis: A 1:100 dilution of this antibody detected Vesicular Glutamate Transporter 2 in adult mouse brain tissue.

Target description

The unconjugated parent antibody (Cat. No. MAB5504) has an observed molecular weight at ~56 kDa

Physical form

Protein A purified
Purified mouse monoclonal IgG1 conjugated to Alexa Fluor 488 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Storage and Stability

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Adult mouse brain tissue

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sami Piirainen et al.
Glia, 69(10), 2459-2473 (2021-06-20)
Microglial activation has been regarded mainly as an exacerbator of stress response, a common symptom in psychiatric disorders. This study aimed to determine whether microglia contribute to adaptive response of the brain and behavior toward stress using a mild and
Keerthana Chithanathan et al.
Biology, 11(6) (2022-06-25)
Anxiety is a known comorbidity and risk factor for conversion to neuroinflammation-mediated dementia in patients with Alzheimer's disease (AD). Here, we investigated if anxiety occurred as an early endophenotype of mutant familial AD (5 × FAD) male mice and the
Keerthana Chithanathan et al.
Frontiers in immunology, 13, 996415-996415 (2022-11-18)
The miR-146 family consists of two microRNAs (miRNAs), miR-146a and miR-146b (miR-146a/b), both of which are known to suppress immune responses in a variety of conditions. Here, we studied how constitutive deficiency of miR-146b (Mir146b-/-) affects lipopolysaccharide (LPS)-induced neuroinflammation in
Ling Yan et al.
Frontiers in cellular neuroscience, 17, 1254923-1254923 (2023-09-29)
Microglia play an important role in the maintenance of brain and behavioral homeostasis. The protective effect of microglial replenishment was reported in neurological diseases, but whether microglial therapy would benefit psychiatric disorders such as schizophrenia has been unclear. As schizophrenia
Friederike Auer et al.
PloS one, 16(3), e0247801-e0247801 (2021-03-03)
Modulation of neuronal excitability is a prominent way of shaping the activity of neuronal networks. Recent studies highlight the role of calcium-activated chloride currents in this context, as they can both increase or decrease excitability. The calcium-activated chloride channel Anoctamin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service