Skip to Content
Merck
All Photos(1)

Key Documents

MAB5300

Sigma-Aldrich

Anti-Dopamine Antibody, clone K56A

clone K56A, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

K56A, monoclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

isotype

IgG

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Dopamine.

The cross-reactivities were determined using an ELISA test by competition experiments with the following compounds:

Compound Cross-reactivity

Dopamine-G-BSA 1

L-DOPA-G-BSA 1/10,000

Tyrosine-G-BSA 1/36,000

Tyramine-G-BSA 1/>50,000

Noradrenaline-G-BSA 1/>50,000

Octopamine-G-BSA 1/>50,000

Adrenaline-G-BSA 1/>50,000

Dopamine 1/>50,000

Abbreviations:

(G) Glutaraldehyde

(BSA) Bovine Serum Albumin

Immunogen

Dopamine-Gluteraldehyde-BSA.

Application

Anti-Dopamine Antibody, clone K56A detects level of Dopamine & has been published & validated for use in IH.
Immunohistochemistry: 1:500-1:2,500 using free floating sections by the PAP technique on rat dopaminergic areas.

Optimal working dilutions must be determined by end user.

PROTOCOL for Dopamine Detection by Immunohisto/cytochemistry. Example for a rat brain.

1. SOLUTIONS TO BE PREPARED - Solution must be prepared as needed.

Solution A: 0.1M cacodylate, 10g/L sodium metabisulfite, pH 6.2.

Solution B: 0.1M cacodylate, 10g/L sodium metabisulfite, 3-5% glutaraldehyde, pH 7.5.

2. RAT PERFUSION - The rat is anaesthetized with sodium pentobarbital or Nembutal and perfused intracardially through the aorta using a pump with Solution A (30 mL): 150-300 mL/min, Solution B (500 mL): 150-300 mL/min.

3. POST FIXATION: 15 to 30 minutes in Solution B, then 4 soft washes in 0.05M Tris with 8.5 g/L sodium metabisulfite, pH 7.5 (Solution C) .

4. TISSUE SECTIONING: Vibratome or cryostat sections can be used.

5. REDUCTION STEP: Sections are reduced with Solution C containing 0.1M sodium borohydride for 10 minutes. The sections are washed 4 times in solution C without sodium borohydride.

6. APPLICATION OF DOPAMINE ANTIBODY: Use a final dilution of 1:2,500-1:10,000 in Solution C containing 0.1% Triton X100 and 2% non-specific serum. Incubate 12 sections per 2 mL diluted antibody overnight, +4°C. Then wash the sections three times for 10 minutes each in Solution C. (Note that the antibody may be usable at a higher dilution. This should be explored to minimize the possibility of high background. Additionally, note that a change in the buffering system as indicated in the protocol may change the background and antibody recognition). The specific reaction is then revealed by PAP procedure.

6. SECOND ANTIBODY: Incubate the sections with a 1:50 to 1:200 dilution of goat anti-rabbit in Solution B containing 1% non-specific serum for either 3 hrs at 20°C or 2 hr at 37°C. Then wash the sections, 3 times, for 10 minutes each with Solution B.

7. PAP: Incubate the sections with the appropriate dilution of peroxidase anti-peroxidase (for free floating method) in Solution B containing 1% non-specific serum for 1-2 hours at 37°C. Then wash sections 3 times for 10 min each in solution B.

8. VISUALIZATION: The antigen-antibody complexes are visualized using DAB-4-HCl (25 mg/100 mL) (or other chromogen) in 0.05M Tris and filtrated; 0.05% hydrogen peroxide is added. Incubate the sections for 10 minutes at room temp. Stop the reaction by transferring the sections to 5 mL 0.05M Tris. Wash tissue with solution D using 2, 10 min washes. Mount sections on chrome-alum coated slides. Dry overnight at 37°C. Rehydrate sections using conventional histological procedures. Coverslip using rapid mounting media.

For research use only; not for use as a diagnostic.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Physical form

Format: Purified
Purified immunoglobulin. Liquid containing 50% glycerol.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanzhong Xu et al.
eNeuro, 4(3) (2017-06-01)
Leptin receptors (LepRs) expressed in the midbrain contribute to the action of leptin on feeding regulation. The midbrain neurons release a variety of neurotransmitters including dopamine (DA), glutamate and GABA. However, which neurotransmitter mediates midbrain leptin action on feeding remains
K Cichewicz et al.
Genes, brain, and behavior, 16(3), 394-403 (2016-10-21)
Dopamine (DA) is a neurotransmitter with conserved behavioral roles between invertebrate and vertebrate animals. In addition to its neural functions, in insects DA is a critical substrate for cuticle pigmentation and hardening. Drosophila tyrosine hydroxylase (DTH) is the rate limiting
Jitte Groothuis et al.
Cell and tissue research, 379(2), 261-273 (2019-08-24)
An extreme reduction in body size has been shown to negatively impact the memory retention level of the parasitic wasp Nasonia vitripennis. In addition, N. vitripennis and Nasonia giraulti, closely related parasitic wasps, differ markedly in the number of conditioning
Emma van der Woude et al.
Brain, behavior and evolution, 89(3), 185-194 (2017-05-10)
Trichogramma evanescens parasitic wasps show large phenotypic plasticity in brain and body size, resulting in a 5-fold difference in brain volume among genetically identical sister wasps. Brain volume scales linearly with body volume in these wasps. This isometric brain scaling
Dimitri Ryczko et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(40), 9759-9770 (2017-09-20)
The mesencephalic locomotor region (MLR) plays a crucial role in locomotor control. In vertebrates, stimulation of the MLR at increasing intensities elicits locomotion of growing speed. This effect has been presumed to result from higher brain inputs activating the MLR

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service