Skip to Content
Merck
All Photos(1)

Documents

MAB3826

Sigma-Aldrich

Anti-Mucin 5B Antibody, clone 19.4E

clone 19.4E, Chemicon®, from mouse

Synonym(s):

MUC5B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

19.4E, monoclonal

species reactivity

monkey, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MUC5B(727897)

General description

Mucins are a family of highly glycosylated proteins that are present on the surface of, and are secreted by, many glandular epithelial cells. Mucins provide a barrier between the luminal membranes of cells in the epithelium and their environment. For instance, in the digestive tract, the mucin barrier protects the epithelial cells from the action of proteolytic enzymes. These high molecular weight glycoproteins consist of a nonglobular protein backbone (apomucin) and many O-linked glycans. Salivary Mucin 5B proteins are some of the largest polypeptides, with molecular weights ranging from 14 - 40 x 103 kDa.

Specificity

Reacts with Mucin 5B (MUC5B). Does not react with human Mucin 5A or 5C.

Immunogen

Un-glycosylated, serine- and threonine-free synthetic peptide from the N-terminus of human MUC5B.

Application

Detect Mucin 5B using this Anti-Mucin 5B Antibody, clone 19.4E validated for use in ELISA, WB, IH.
Research Category
Apoptosis & Cancer
Research Sub Category
Tumor Markers
Western blot; Bands in the high molecular weight range of 200kDa or above are expected for highly glycosylated proteins.

Immunohistchemistry: Paraffin sections. Deglycosylating the tissue is critical to expose epitope. There are a number of deglycosylation protocols available and a discussion of these related to Mucin can be found in Cao, Y. et al. (1997)(see references).

ELISA

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin. Liquid in PBS containing 0.1% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service