MAB2642
Anti-Fibrillin-2 Antibody, clone 48
clone 48, from mouse
Synonym(s):
fibrillin 2, congenital contractural arachnodactyly, Fibrillin 5
About This Item
Recommended Products
biological source
mouse
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
48, monoclonal
species reactivity
human
technique(s)
ELISA: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... FBN2(2201)
General description
Specificity
Immunogen
Application
Immunofluorescence Analysis: A previous lot was used by an independent laboratory in IF (Charbonneau, 2003).
Quality
Immunohistochemistry Analysis: 1:50 dilution of this antibody detected Fibrillin-2 in paraffin-embedded tissue sections of liver cancer and papillary serous carcinoma.
Target description
Physical form
Analysis Note
Liver cancer and papillary serous carcinoma tissue
Other Notes
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service