Skip to Content
Merck
All Photos(1)

Documents

MAB1592-C

Sigma-Aldrich

Anti-Neurofilament NF-H Antibody, phosphorylated Antibody, clone NP1

clone NP1, from mouse

Synonym(s):

Neurofilament heavy polypeptide, NF-H, 200 kDa neurofilament protein, Neurofilament triplet H protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

NP1, monoclonal

species reactivity

canine, mouse, human, rat

species reactivity (predicted by homology)

chicken (based on 100% sequence homology), bovine (based on 100% sequence homology), porcine (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

phosphorylation (not specified)

Gene Information

bovine ... Nefh(528842)
chicken ... Nefh(417020)
dog ... Nefh(442940)
human ... NEFH(4744)
mouse ... Nefh(380684)
pig ... Nefh(100156492)
rat ... Nefh(24587)

General description

Neurofilament heavy polypeptide (UniProt: P12036; also known as NF-H, 200 kDa neurofilament protein, Neurofilament triplet H protein) is encoded by the NEFH (also known as KIAA0845, NFH) gene (Gene ID: 4744) in human. Neurofilaments (~ 10 nm in diameter) are intermediate filaments that serve as major elements of the cytoskeleton supporting the axon cytoplasm. They are the most abundant fibrillar components of the axon, being on average 3-10 times more frequent than axonal microtubules. They are built from three intertwined protofibrils, which are themselves composed of two tetrameric protofilament complexes of monomeric proteins. The neurofilament triplet proteins (68/70, 160, and 200 kDa) occur in both the central and peripheral nervous system and are usually neuron specific. The 68/70 kDa NF-L protein can self-assemble into a filamentous structure, however the 160 kDa NF-M and 200 kDa NF-H proteins require the presence of the 68/70 kDa NF-L protein to co-assemble. Antibodies to NF-H are useful for identifying neuronal cells and their processes in tissue sections and in tissue culture. NF-H antibodies are helpful in the identification of neurofilament accumulations seen in many neurological diseases, such as Amyotrophic lateral sclerosis (ALS) and Alzheimer′s disease. The purified immunogen used to generate this antibody was the very heavily phosphorylated form of NF-H and this antibody binds only this heavily phosphorylated form. Similar, but far fewer phosphorylation sites are also found in NF-M. Hence, this antibody displays some reactivity with NF-M. Enzymatic phosphorylation of porcine and bovine neurofilaments is shown to result in loss of binding.

Specificity

Clone NP1 detects heavily phosphorylated form of Neurofilament H. Displays weaker reactivity with phosphorylated Neurfilament -M.

Immunogen

Native Neurofilament H purified from pig spinal cord.

Application

Anti-Neurofilament NF-H, phosphorylated, clone NP1, Cat. No. MAB1592-C, is a highly specific mouse monoclonal antibody that targets Neurofilament heavy polypeptide and has been tested for use in ELISA, Immunohistochemistry (Paraffin), and Western Blotting.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected Neurofilament NF-H in human ovary tissue.

Immunohistochemistry Analysis: A representative lot detected Neurofilament NF-H in Immunohistochemistry applications (Benson, D.L., et. al. (1996). J Neurocytol. 25(3):181-96; Boylan, K., et. al. (2009). J Neurochem. 111(5):1182-91).

Western Blotting Analysis: A representative lot detected Neurofilament NF-H in Western Blotting applications (Benson, D.L., et. al. (1996). J Neurocytol. 25(3):181-96; Boylan, K., et. al. (2009). J Neurochem. 111(5):1182-91).

ELISA Analysis: A representative lot detected Neurofilament NF-H in ELISA applications (Boylan, K., et. al. (2009). J Neurochem. 111(5):1182-91; Toedebusch, C.M., et. al. (2017). J Vet Intem Med. 31(2):513-520; Mashita, T., et. al. (2015). J Vet Med Sci. 77(4):433-8).
Research Category
Neuroscience

Quality

Evaluated by Immunohistochemistry in human cerebral cortex tissue.

Immunohistochemistry Analysis: A 1:50 dilution of this antibody detected Neurofilament NF-H in human cerebral cortex tissue.

Target description

112.48 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibody IgG1 in PBS with 5 mM sodium azide and 50% glycerol.

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service