Skip to Content
Merck
All Photos(1)

Documents

LSKMAGKP02

Millipore

PureProteome Kappa Ig Binder Magnetic Beads

The PureProteome Kappa Ig Binder Magnetic Bead System is a powerful system that helps researchers purify all Immunoglobulins with high specificity.

Synonym(s):

Kappa Ig Binder Magnetic Beads, PureProteome Magnetic Beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116133
eCl@ss:
32160405
NACRES:
NA.56

packaging

pkg of 2 mL

manufacturer/tradename

PureProteome

technique(s)

protein purification: suitable (antibody)

particle size

10 μm

capacity

≥2.2 μg/μL, slurry binding capacity (kappa IgG)

shipped in

wet ice

storage temp.

2-8°C

General description

The PureProteome Kappa Ig Binder Magnetic Beads have been developed with a specialized antibody ligand that specifically targets the human kappa light chain. These magnetic beads offer a fast, scalable, and reliable method for capturing human antibodies or antibody fragments that possess kappa light chains. With high specificity, the Kappa Magnetic Beads bind effectively to the constant region of the kappa light chain found in human immunoglobulins.

Application

PureProteome Kappa Ig Binder Magnetic Beads are suitable:
  • for antibody purification
  • for protein purification
  • to perform coimmunoprecipitation of immune complexes (ICs) in mouse serum to capture host antibody interactions with the circulatory antigens of Trichinella spiralis
  • to capture the immune complexes in the sera of microfilaremic subjects

Features and Benefits

  • More binding sites for proteins due to advanced chemistry and high surface area.
  • Consistent results with no sample loss.
  • Fast processing time.
  • These magnetic beads are more affordable compared to competing alternatives.

Legal Information

PureProteome is a trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunome and immune complex-forming components of Brugia malayi identified by microfilaremic human sera
Reamtong O, et al.
Experimental Parasitology (2019)
Identification and profiling of Trichinella spiralis circulating antigens and proteins in sera of mice with trichinellosis
Charin T, et al.
PLoS ONE (2022)
Identification and profiling of Trichinella spiralis circulating antigens and proteins in sera of mice with trichinellosis
Charin T, et al.
Testing (2022)

Articles

PureProteome™ Protein A and G Magnetic beads provide a rapid and reproducible means to purify immunoglobulins (IgG) using the KingFisher Duo particle processor.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service