Skip to Content
Merck
All Photos(1)

Documents

HX0645

Supelco

Hydrogen Peroxide Solution 3% W/V

stabilized TITRISTAR

Synonym(s):

Peroxide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51473503

Agency

traceable to NIST

form

liquid

shipped in

wet ice

storage temp.

2-8°C

Specifications

Tolerance: 3.0-3.5% W/V

Analysis Note

Expiration date: Exp 12 months from mfg date
Arsenic (As): 0.5ppm max
Assay: 3.0% min
Chloride (Cl): 0.001% max
Color (APHA): 20 max
Evaporation residue: 0.02% max
Form: Clear liquid
Free acid (as H2SO4): 0.01% max
Heavy metals (as Pb): 1ppm max
Iron (Fe): 5ppm max
Nitrogen compounds (as N): 0.005% max
Phosphate (PO4): 0.003% max
Preservative (acetanilide): 0.02% max
Sulfate (SO4): 0.005% max

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service