Skip to Content
Merck
All Photos(2)

Key Documents

HTS111M

Sigma-Aldrich

ChemiSCREEN Membrane Preparation Recombinant Human 5-HT6 Serotonin Receptor

Human 5-HT6 GPCR membrane preparation for Radioligand binding Assays & GTPγS binding.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.84

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Full-length human HTR6 cDNA encoding the 5-HT6 Serotonin Receptor
The neurotransmitter serotonin/5-hydroxytryptamine (5-HT) regulates a wide variety of neurological functions. A family of 13 receptors (12 GPCRs and one ion channel) mediate the effects of serotonin. The serotonin receptor 5-HT6 is a Gs coupled receptor expressed solely in the CNS, primarily in the limbic and cortical regions. 5-HT6 appears to play a role in memory and learning, obesity, psychosis, anxiety and epilepsy (Woolley et al., 2004; Fisas et al., 2006). In particular, a 5-HT6-selective agonist caused significant weight loss in a rat model of diet-induced obesity. Millipore′s 5-HT6 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of agonists and antagonists of 5-HT6. The membrane preparations exhibit a Kd of 0.44 nM for [125I]-SB258585. With 0.25nM [125I]-SB258585, 5 μg/well 5-HT6 Membrane Prep typically yields greater than 6-fold signal-to-background ratio.

Application

Radioligand binding assay and GTPγS binding.

Biochem/physiol Actions

GPCR Class: A
Protein Target: 5-HT6
Target Sub-Family: Serotonin

Quality

Table 1. Signal:background and specific binding values obtained in a competition binding assay with varying amounts of 5-HT6 Receptor membrane prep.
5 µg/well 10 µg/well
Signal:Background 12.8 12.3
Specific Binding (cpm) 10192 12675


SPECIFICATIONS: 1 unit = 5 µg membrane preparation
Bmax for [125I]-SB258585 binding: 6.1 pmol/mg protein
Kd for [125I]-SB258585 binding: ~0.44 nM

Specifications

Inucbation Conditions
RECOMMENDED ASSAY CONDITIONS: Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, an FC 96-well harvest plate (Millipore cat. # MAHF C1H) is coated with 0.33% polyethyleneimine for 30 min, then washed with the binding buffer. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.

Binding buffer: 20 mM HEPES, 3 mM MgCl2, 2 mM Ascorbic acid, pH 7.4, filtered and stored at 4°C

Radioligand: [125I]-SB258585 (Perkin Elmer #:NEX424)

Wash Buffer: same as the binding buffer.

One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 6-fold signal:background with 125I labeled SB258585 at 0.25 nM

Physical form

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA with no preservatives.
Packaging method: Membranes protein were adjusted to the indicated concentration in packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at –70°C up to expiration date indicated on the label. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chronic 5-HT6 receptor modulation by E-6837 induces hypophagia and sustained weight loss in diet-induced obese rats.
Fisas, Angels, et al.
British Journal of Pharmacology, 148, 973-983 (2006)
5-ht6 receptors.
Woolley, Marie L, et al.
Current Drug Targets. CNS and Neurological Disorders, 3, 59-79 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service