Skip to Content
Merck
All Photos(2)

Key Documents

HTS092M

Sigma-Aldrich

ChemiSCREEN EP3 Prostanoid Receptor Membrane Preparation

Human EP3 GPCR membrane preparation for Radioligand binding Assays & GTPgammaS binding.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32161000
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

human

Quality Level

recombinant

expressed in Chem-1 cells

manufacturer/tradename

ChemiScreen
Chemicon®

technique(s)

ligand binding assay: suitable (GTPγS)
radioligand binding assay (RLBA): suitable

UniProt accession no.

shipped in

dry ice

Gene Information

human ... PTGER3(5733)

General description

Full-length human PTGER3 cDNA encoding splice variant 6 of EP3
Prostanoids bind to a family of 8 GPCRs to exert their biological effects (Narumiya and FitzGerald, 2001). The prostaglandin PGE2 causes pain, vasodilation, immunosuppression of T cells, bone resorption and promotion of carcinogenesis. Four related GPCRs, EP1, EP2, EP3 and EP4, each bind to PGE2, but the different G protein coupling status of each receptor leads to distinct biological effects. Further diversity is generated by alternative splicing; the human gene for EP3 generates 9 alternatively spliced mRNAs encoding 8 isoforms of EP3 (Kotani et al., 1997). These isoforms of EP3 vary in sequence at their C-termini, and differ in their ability to couple to Gs, Gq or Gi (Kotani et al., 1995). EP3 is required for fever induced by pyrogens, a response long attributed to prostaglandins by the antipyretic action of aspirin and other COX inhibitors (Ushikubi et al., 1998). Chemicon′s EP3 membrane preparations are crude membrane preparations made from our proprietary stable recombinant cell lines to ensure high-level of GPCR surface expression; thus, they are ideal HTS tools for screening of antagonists of EP3 interactions and its ligands. The membrane preparations exhibit a Kd of 1.56 nM for [3H]-PGE2. With 1.5 nM [3H]-PGE2, 5 µg/well and 10 µg/well EP3 Membrane Prep typically yield greater than 10 and 20-fold signal-to-background ratios, respectively.

Application

Radioligand binding assay and GTPS binding.

Biochem/physiol Actions

GPCR Class: A
Protein Target: EP3
Target Sub-Family: Prostanoid

Quality

Signal:background and specific binding values obtained in a competition binding assay with varying amounts of EP3 membrane prep:
10 µg/well5 µg/well
Signal:Background 38 21
Specific Binding (cpm) 6,386 4,053


Specifications:
1 unit = 10 μg
Bmax: 15.0 pmol/mg
Kd: 1.56 nM

Specifications

Inucbation Conditions
Membranes are mixed with radioactive ligand and unlabeled competitor (see Figures 1 and 2 for concentrations tested) in binding buffer in a nonbinding 96-well plate, and incubated for 1-2 h. Prior to filtration, a GF/C 96-well filter plate is coated with 0.33% polyethyleneimine for 30 min, then washed with 50mM HEPES, pH 7.4, 0.5% BSA. Binding reaction is transferred to the filter plate, and washed 3 times (1 mL per well per wash) with Wash Buffer. The plate is dried and counted.
Binding buffer: 50 mM Tris, pH 7.4, 10 mM MgCl2, 1 mM EDTA, filtered and stored at 4°C
Radioligand: [3H] PGE2 (Perkin Elmer#:NET-428 )
Wash Buffer: 50 mM Tris, pH 7.4 filtered and stored at 4°C.

Physical form

One package contains enough membranes for at least 200 assays (units), where a unit is the amount of membrane that will yield greater than 10-fold signal:background with 3H-labeled PGE2 at 1.5 nM

Liquid in packaging buffer: 50 mM Tris pH 7.4, 10% glycerol and 1% BSA no preservatives.
Packaging method: Membranes protein were adjusted to 0.5 mg/ml in 1 ml packaging buffer, rapidly frozen, and stored at -80°C.

Storage and Stability

Maintain frozen at -70°C for up to 2 years. Do not freeze and thaw.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Ushikubi et al.
Nature, 395(6699), 281-284 (1998-09-29)
Fever, a hallmark of disease, is elicited by exogenous pyrogens, that is, cellular components, such as lipopolysaccharide (LPS), of infectious organisms, as well as by non-infectious inflammatory insults. Both stimulate the production of cytokines, such as interleukin (IL)-1beta, that act
M Kotani et al.
Molecular pharmacology, 48(5), 869-879 (1995-11-01)
Five distinct cDNA clones encoding four different isoforms of human prostaglandin (PG) E receptor EP3 subtype were isolated from a human kidney cDNA library. Two cDNA clones differed only in their 3'-untranslated regions. The four isoforms, tentatively named EP3-I, EP3-II
M Kotani et al.
Genomics, 40(3), 425-434 (1997-03-15)
Prostaglandin EP3 receptor subtype is a seven-membrane-spanning protein with multiple C-terminal tails generated by alternative mRNA splicing. We report here the structural organization of the human EP3 gene (PTGER3). The human EP3 gene spanned more than 80 kb and was
Genetic and pharmacological analysis of prostanoid receptor function.
S Narumiya et al.
The Journal of clinical investigation, 108(1), 25-30 (2001-07-04)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service