Skip to Content
Merck
All Photos(1)

Key Documents

FXWXM20K0001

Millipore

Mobius® Chrom 20 Flexware® Kit

1mm Post

Synonym(s):

Mobius® Chrom 20 Flexware® Kit (1mm Post)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

Quality Level

product line

Mobius®

packaging

pkg of 1 packs

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Legal Information

FLEXWARE is a registered trademark of Merck KGaA, Darmstadt, Germany
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service