Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB323PE

Sigma-Aldrich

Milli-Mark® Anti-Phosphotyrosine-PE Antibody, recombinant clone 4G10®

clone 4G10, Milli-Mark®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4G10, monoclonal

species reactivity

vertebrates, human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG2bκ

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr)

Gene Information

human ... PID1(55022)

Related Categories

General description

Some of the tyrosine residues can be tagged with a phosphate group by protein kinases and in its phosphorylated state, it is referred to as phosphotyrosine. Tyrosine phosphorylation is considered as one of the key steps in signal transduction and regulation of enzymatic activity.
The advent of anti-phosphotyrosine antibodies is one of the significant events in signal transduction research. Before the availability of anti-phosphotyrosine antibodies, tyrosine phosphorylation of proteins and enzymes was investigated through hazardous and time-consuming radioactive experiments. Anti-phosphotyrosine antibodies are commonly used in western blots after the targeted proteins have been immunoprecipitated to measure the tyrosyne phosphorylation of the proteins. Anti-phosphotyrosine antibodies are also directly used on cell lysate to examine the overall change of tyrosine phosphorylation level in reponse to various treatments.

Specificity

Antibody recognizes tyrosine-phosphorylated proteins from all species
Expected to detect tyrosyne phosphorylated proteins in all species.

Application

Milli-Mark Anti-Phosphotyrosine-PE Antibody, recombinant clone 4G10 is an antibody against Phosphotyrosine clone 4G10 for use in FC.

Quality

Evaluated by flow cytometry using EGF treated versus untreated A431 cells

Target description

Dependent upon the molecular weight of the tyrosine phosphorylated protein being detected.

Physical form

Purified mouse monoclonal IgG2bk conjugated to PE in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Analysis Note

Control
Treated versus untreated A431 cells

Legal Information

4G10 is a registered trademark of Upstate Group, Inc.
MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neel H Shah et al.
eLife, 7 (2018-03-17)
The specificity of tyrosine kinases is attributed predominantly to localization effects dictated by non-catalytic domains. We developed a method to profile the specificities of tyrosine kinases by combining bacterial surface-display of peptide libraries with next-generation sequencing. Using this, we showed
Veronika Horkova et al.
Cell reports, 30(5), 1504-1514 (2020-02-06)
Overtly self-reactive T cells are removed during thymic selection. However, it has been recently established that T cell self-reactivity promotes protective immune responses. Apparently, the level of self-reactivity of mature T cells must be tightly balanced. Our mathematical model and experimental data

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service