EZHABG504
EZ-Pak® Membrane Filter
mixed cellulose esters (MCE), pore size 0.45 μm, filter diam. 50 mm, black filter, gridded filter, General microbiological analysis
Synonym(s):
Membrane filters, Semi-permeable membranes
About This Item
Recommended Products
material
black filter
gridded filter
mixed cellulose esters (MCE)
Quality Level
Agency
ISO 7704
sterility
sterile; ethylene oxide treated
feature
easily wetted with water
hydrophilic
individually wrapped
manufacturer/tradename
EZ-Pak®
filter diam.
50 mm
matrix
MF-Millipore™
pore size
0.45 μm pore size
input
sample type bottled water
sample type beer
sample type soft drink
sample type cider
sample type food(s)
sample type sport drink
sample type wine
sample type pharmaceutical(s)
sample type beverage(s)
sample(s) (environmental)
sample type cosmetics
application(s)
bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
pharmaceutical
shipped in
ambient
Related Categories
General description
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Sol. 1
Storage Class Code
4.1B - Flammable solid hazardous materials
WGK
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service