Skip to Content
Merck
All Photos(1)

Key Documents

EX0242

Supelco

Ethyl Acetate Biosynthesis

Biosynthesis OmniSolv®

Synonym(s):

Ethyl acetate, Acetic Acid Ethyl Ester, EtOAc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOC2H5
CAS Number:
Molecular Weight:
88.11
Beilstein:
506104
MDL number:
UNSPSC Code:
12352108
EC Index Number:
205-500-4
bp:
76.5-77.5 °C (lit.)
vapor pressure:
73 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3 (20 °C, vs air)

Quality Level

vapor pressure

73 mmHg ( 20 °C)

product line

OmniSolv®

Assay

≥99.9% (GC)

form

liquid

autoignition temp.

801 °F

expl. lim.

2.2-11.5 %, 38 °F

impurities

≤0.8 μeq/g Titrable acid
≤100 ppm Water (H2O)
≤250 ppt Fluorescence (as quinine base)

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.3720 (lit.)

bp

76.5-77.5 °C (lit.)

mp

−84 °C (lit.)

transition temp

flash point -4 °C

solubility

85.3 g/L

density

0.902 g/mL at 25 °C (lit.)

UV cutoff

≤255 nm

UV absorption

λ: 255 nm Amax: ≤1.00
λ: 257 nm Amax: ≤0.35
λ: 263 nm Amax: ≤0.06
λ: 275 nm Amax: ≤0.02
λ: 300 nm Amax: ≤0.01

format

neat

shipped in

ambient

storage temp.

room temp

SMILES string

CCOC(C)=O

InChI

1S/C4H8O2/c1-3-6-4(2)5/h3H2,1-2H3

InChI key

XEKOWRVHYACXOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Biosynthesis OmniSolv®

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (GC)99.9% minColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)250ppt maxFormClear liquidInfrared SpectrumConforms to standardTitrable acid0.8µeq/g maxUV Abs. at 255 nm1.00AU maxUV Abs. at 257 nm0.35AU maxUV Abs. at 263 nm0.06AU maxUV Abs. at 275 nm0.02AU maxUV Abs. at 300 nm0.01AU maxUV Cut-off255nm maxWater (H2O)100ppm max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The 4L M-Bottle provides safe handling of solvents due to an innovative solvent bottle design with sealing technology to eliminate leaks and reduce risks in solvent dispensing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service