Skip to Content
Merck
All Photos(1)

Key Documents

EGARDT1X1

Milli-Q

E-Gard® Purification Pack

Designed to remove trace of ionic contaminants affecting specific applications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
NA.21

Quality Level

feature

removes trace contaminants

packaging

pkg of 1 unit

manufacturer/tradename

E-Gard®

technique(s)

ion chromatography: suitable

H

32 cm (12.6 in.)

compatibility

for use with Elix® Advantage

shipped in

ambient

General description

E-Gard® Purification Pack has been designed to improve pure water quality by removing trace of ionic contaminants.

Features and Benefits

  • E-Gard® Purification Pack includes a combination of purification media for effective removal of trace contaminants affecting water purity.
  • Constructed from polypropylene and polyethylene which are suitable to produce pure and ultrapure water and have been qualified for low organics and ionic extractable.
  • Ion exchange resin cartridge

Legal Information

E-GARD is a registered trademark of Merck KGaA, Darmstadt, Germany
ELIX is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service