Skip to Content
Merck
All Photos(1)

Documents

EDSPAK001

Milli-Q

EDS-Pak® Polisher

Production of endocrine disruptor-free water

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104212
eCl@ss:
33050190
NACRES:
NA.21

material

carbon cartridge (activated)
polyethylene (Luer Plug)
polyethylene connector (Vent Filter)
polypropylene housing

Quality Level

feature

removes endocrine disruptors such as phthalate esters, Bisphenol-A and Nonylphenol

packaging

pkg of 1 unit

manufacturer/tradename

EDS-Pak®

parameter

2 L/min max. flow rate
2 bar pressure
5 °C temp. range (30 °C)

L

12.7 cm

impurities

<5 ppb TOC

compatibility

for use with Direct-Q® UV
for use with Direct-Q®
for use with Milli-Q® Advantage A10
for use with Milli-Q® Direct
for use with Milli-Q® Reference
for use with Simplicity® UV
for use with Simplicity®
for use with Synergy® UV
for use with Synergy®

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

EDS-Pak® Polisher is an activated carbon-based cartridge to be connected at the outlet of ultrapure water equipment, at the outlet of the Q-POD® dispenser of the Milli-Q® Integral and Advantage A10 for the best results. EDS-Pak® cartridge media, it′s housing, and connector materials are validated for the removal of several endocrine disrupters.
EDS-Pak® polisher can be installed on all Type 1 water purification systems that deliver ultrapure water with TOC < 5 ppb, including the Milli-Q®, Synergy®, Simplicity® and Direct-Q® systems.

Application

The EDS-Pak® polisher delivers endocrine disruptor-free water suitable for the preparation of blanks, reagents, and standard solutions used for the analyses of molecules such as Bisphenol A, phthalates and nonylphenol by GC-MS or LC-MS.

Features and Benefits

  • Point-of-use polisher for use with Milli-Q® and other Type 1 water purification systems.
  • Final purification step provides endocrine disrupter-free ultrapure water at a high flow rate.
  • Delivers a minimum of 300 L of EDS-free ultrapure water.
  • EDS-Pak® cartridge contains a specific type of activated carbon validated for efficient removal of endocrine disrupters (EDS) mainly bisphenol A, diethyl phthalate and di-n-butyl phthalate.
  • Each EDS-Pak® cartridge is delivered hermetically sealed, with its Certificate of Quality.

Other Notes

Before use, the EDS-Pak® unit must be conditioned with methanol to ensure that all binding surfaces are accessible to endocrine disrupters. Following this step, the cartridge should then be flushed with ultrapure water to remove any excess methanol. Once conditioned, the EDS-Pak® unit is warranted to deliver up to 300 L of EDS-free water when fed by ultrapure water with a TOC level < 5 ppb at a flow rate between 0.5 – 2 L/min.

A 50 mL glass syringe is required to condition the EDS-Pak® cartridge with methanol. This syringe is not provided.

Legal Information

DIRECT-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
EDS-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Q-POD is a registered trademark of Merck KGaA, Darmstadt, Germany
SIMPLICITY is a registered trademark of Merck KGaA, Darmstadt, Germany
SYNERGY is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service