CDUFHF05K
Pyrogard® 5000 Ultrafiltration Cartridge
For use with Milli-Q® Biocel and Synthesis systems; to produce pyrogen- and nuclease-free water
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Quality Level
feature
hydrophobic
packaging
pkg of 1 unit
manufacturer/tradename
Pyrogard®
parameter
1.0 L/min flow rate
resistivity
18.2 MΩ-cm (at 25 degree celcius)
impurities
<0.001 EU/mL Pyrogen Level
fitting
6 mm tubing (UF cartridge reject tubing)
compatibility
for use with Milli-Q® Biocel
for use with Milli-Q® Synthesis
shipped in
ambient
Related Categories
General description
Pyrogard® 5000 Ultrafiltration Cartridge (UF) is a hollow-fiber in-line ultrafilter used with Milli-Q® Biocel and Synthesis, to produce pyrogen- and nuclease-free ultrapure water.
Application
Water filtered through this UF cartridge is recommended for experiments and analyses where nuclease-free and/or endotoxin-free water is needed.
Features and Benefits
- Comprises polysulfone hollow fiber membranes in an acrylonitrile-butadiene-styrene housing with polyurethane potting compound. Initially contains 1.5 vol. % hydrogen peroxide storage solution.
- Purification technology involves the use of ion exchange, activated carbon membrane.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
PYROGARD is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service