Skip to Content
Merck
All Photos(1)

Key Documents

BX1626

Supelco

Buffer Variety Pack, Kit

Synonym(s):

buffers

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161703

grade

analytical standard

Agency

traceable to NIST

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

shipped in

ambient

storage temp.

15-25°C

General description

The buffer variety pack contains two solutions each of pH buffer 4 (red, BX1628), pH buffer 7 (yellow, 1632), and pH 10 buffer (blue, 1641).

Application

The buffer solutions are appropriate for use in daily analytical procedures involving the calibration of pH meters and maintaining pH of solutions.

Features and Benefits

  • Traceable to NIST standards
  • Ready-to-use solutions n available in a sealed bottle with convenient packaging to protect against contamination
  • Accompanied with a certificate of analysis (CoA) for an effective audit
  • The expiry date of the product can be found in the CoA
  • Calculated uncertainty value is for the duration of transportation and storage

Components

Pack: 2 x 500mL each pH 4 Red (BX1628), pH 7 Yellow (BX1632) and pH 10 Blue (BX1641).

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service