AX1222
Ammonium Acetate
HPLC, Meets ACS Specifications
Synonym(s):
Ammonium Acetate, buffers
About This Item
Recommended Products
Quality Level
Assay
≥98.0%
form
crystals
technique(s)
HPLC: suitable
impurities
≤0.005% Insoluble matter
ign. residue
≤0.01%
pH
6.7-7.3 (25 °C, 50 g/L in H2O)
mp
114 °C
solubility
1480 g/L
anion traces
chloride (Cl-): ≤5 ppm
nitrate (NO3-): ≤0.001%
sulfate (SO42-): ≤0.001%
cation traces
Fe: ≤5 ppm
heavy metals (as Pb): ≤5 ppm
UV absorption
λ: 254 nm Amax: ≤0.02
λ: 280 nm Amax: ≤0.01
λ: 350 nm Amax: ≤0.01
shipped in
ambient
storage temp.
2-8°C
InChI
1S/C2H4O2.H3N/c1-2(3)4;/h1H3,(H,3,4);1H3
InChI key
USFZMSVCRYTOJT-UHFFFAOYSA-N
Application
Analysis Note
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service