Skip to Content
Merck
All Photos(1)

Documents

AX0445

Supelco

Reagent Alcohol

89.5-91.5% ethanol basis, Reagent, OmniSolv®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6O
CAS Number:
Molecular Weight:
46.07
MDL number:
UNSPSC Code:
12352104

product name

Alcohol, Reagent, OmniSolv®

Quality Level

Assay

89.5-91.5% ethanol basis

form

liquid

composition

isopropyl alcohol, 4.0-6.0%
methanol, 3.5-5.5%

impurities

≤0.04% Water (H2O)
≤0.04 μeq/g Titrable base
≤0.3 μeq/g Titrable acid

evapn. residue

≤4 ppm

color

APHA: ≤10

transition temp

flash point 12 °C

UV cutoff

≤205 nm

UV absorption

λ: 205 nm Amax: ≤1.00
λ: 210 nm Amax: ≤0.65
λ: 220 nm Amax: ≤0.35
λ: 230 nm Amax: ≤0.20
λ: 250 nm Amax: ≤0.04
λ: 270 nm Amax: ≤0.01
λ: 300 nm Amax: ≤0.005

shipped in

ambient

storage temp.

room temp

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent, OmniSolv®

Physical form

To each 100 parts of SD 3A Alcohol are added 5 parts of Isopropyl Alcohol

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (Ethanol)89.5-91.5%Assay (Isopropyl alcohol)4.0-6.0%Assay (Methanol)3.5-5.5%Color (APHA)10 maxEvaporation residue4ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidInfrared SpectrumConforms to standardTitrable acid0.3µeq/g maxTitrable base0.04µeq/g maxUV Abs. at 205 nm1.00AU maxUV Abs. at 210 nm0.65AU maxUV Abs. at 220 nm0.35AU maxUV Abs. at 230 nm0.20AU maxUV Abs. at 250 nm0.04AU maxUV Abs. at 270 nm0.01AU maxUV Abs. at 300 nm0.005AU maxUV Cut-off205nm maxWater (H2O)0.04% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service