AX0125
Acetone
Histology
Synonym(s):
Dimethyl Ketone, 2-Propanone
About This Item
Recommended Products
grade
histological grade
Quality Level
vapor density
2 (vs air)
vapor pressure
184 mmHg ( 20 °C)
Assay
≥99% (GC)
form
liquid
expl. lim.
13.2 %
evapn. residue
≤0.001%
color
APHA: ≤10
refractive index
n20/D 1.359 (lit.)
pH
5-6 (20 °C, 395 g/L in H2O)
bp
56 °C/760 mmHg (lit.)
mp
−94 °C (lit.)
transition temp
flash point <-20 °C
density
0.791 g/mL at 25 °C (lit.)
application(s)
histology
format
neat
shipped in
ambient
storage temp.
room temp
SMILES string
CC(C)=O
InChI
1S/C3H6O/c1-3(2)4/h1-2H3
InChI key
CSCPPACGZOOCGX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Analysis Note
Color (APHA): 10 max
Evaporation residue: 0.001% max
Form: Clear liquid
Infrared Spectrum: Conforms to standard
Readily oxidizable substances: To pass test
Solubility: To pass test
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
1.4 °F - closed cup
Flash Point(C)
-17.0 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service