Skip to Content
Merck
All Photos(1)

Documents

AP500P

Sigma-Aldrich

Goat Anti-Mouse IgM µ chain Antibody, HRP conjugate, Species Adsorbed

Chemicon®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

peroxidase conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

IgM constitutes about 10% of serum immunoglobulins. IgM antibody is prominent in early immune responses to most antigens and predominates in certain antibody responses such as ′natural′ blood group antibodies. IgM (with IgD) is the major immunoglobulin expressed on the surface of B cells. The gene for the mu constant region contains four domains separated by short intervening sequences.

Specificity

Reacts with heavy chain of mouse IgM. Absorbed for mouse IgG1, IgG2a, IgG2b, IgG3 and IgA, pooled human sera and purified human paraproteins. Minimal cross reactivity with human immunoglobulins. Reactivity with other species has not been determined.
The antibody reacts with the heavy chain of mouse IgM as demonstrated by ELISA and flow cytometry. Minimal cross reactivity with human immunoglobulins.

Immunogen

Epitope: IgM µ chain
Prepared from purified mouse IgM µ chain. Pooled antisera from goats hyperimmunized with mouse IgM paraproteins.

Application

Detect Mouse IgM µ chain using this Goat anti-Mouse IgM µ chain Antibody, HRP conjugate, Species Adsorbed validated for use in ELISA.
ELISA: 1:4,000-1:8,000.

Optimal working dilutions must be determined by the end user.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling

Fragment Specific Secondary Antibodies

Physical form

Affinity Chromatography
Purified by affinity chromatography on mouse IgM covalently linked to agarose. Liquid in 50% glycerol/50% PBS, pH 7.4.

Storage and Stability

Maintain refrigerated at 2°-8°C under sterile conditions for up to twelve months from date of receipt. For long term storage, aliquot and store at -20°C. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irati Garmendia et al.
Cancer immunology research, 11(4), 530-545 (2023-03-09)
One billion people worldwide get flu every year, including patients with non-small cell lung cancer (NSCLC). However, the impact of acute influenza A virus (IAV) infection on the composition of the tumor microenvironment (TME) and the clinical outcome of patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service