Skip to Content
Merck
All Photos(1)

Key Documents

AG774

Sigma-Aldrich

Orexin-A, control peptide for AB3098

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.28
Pricing and availability is not currently available.

biological source

mouse

Quality Level

form

liquid

manufacturer/tradename

Chemicon®

technique(s)

inhibition assay: suitable (peptide)

NCBI accession no.

UniProt accession no.

General description

18 amino acid peptide from the amino terminus of mouse Orexin-A.
Orexin-A control peptide.

Several peptides associated with feeding behavior have been reported recently. Orexins (Orexin-A and Orexin-B) are a family of hypothalamic neuropeptides selectively expressed in the hypothalamus (1). Orexin-A and Orexin-B are derived from the same precursor (Prepro-orexin) by proteolytic cleavage. Prepro-orexin is an 130 aa peptide with a putative 33 aa secretory sequence, a hydrophobic core followed by residues with small polar side chains. The expression was detected in brain and to a small extent in testis (1). These neuropeptides bind and activate two closely related Orexin receptors--G protein coupled receptors (GPCRs) OX1R and OX2R.

Application

Blocking peptide for AB3098.

Optimal working concentration must be determined by the end user.

Physical form

Liquid in PBS, pH 7.5

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service