AC464
NovaSeptum® SURe Accurate Volume Syringe Sampling Unit
Manifold (5-valve) with AseptiQuik® S connector, sterile; γ-irradiated, volume 5 mL (20 mL)
Synonym(s):
AV syringe, Accurate Volume syringe, syringe
About This Item
Recommended Products
material
polycarbonate connector
polycarbonate container
polypropylene body
silicone tubing
Quality Level
sterility
sterile; γ-irradiated
product line
NovaSeptum® SURe
storage condition
room temperature
parameter
0.50 bar max. pressure (7.25 psi)
4-40 °C temp. range (39-104 °F)
volume
5 mL (20 mL)
fitting
inlet connection (Silicone)
Luer-Lok® outlet connection (3-piece)
outlet connection (containing a male, female and an injection site)
application(s)
sterile sampling
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
This product comes as a manifolded unit with accurate volume syringes and connects via AseptiQuik® S connector. The syringes enable sampling for small, accurate volumes, which is particularly important for sampling of high value products. The closed design ensures your sample will be isolated from point of sample to analysis, reducing the risk of losing valuable product and maintaining the integrity of your fluid samples.
Application
Features and Benefits
- Product design ensures process integrity and a representative sample from single-use manufacturing processes
- Industry-standard aseptic connection and disconnection for easy integration into your single-use process
- Other container options available: bags, bottles, syringes, and conical tubes
- Stocked and ready-to-ship for fast delivery
Packaging
- 5 units per box (5*5mL)
- 4 units per box (5*20mL)
Preparation Note
Analysis Note
Other Notes
Legal Information
Not finding the right product?
Try our Product Selector Tool.
also commonly purchased with this product
related product
required but not provided
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service