Skip to Content
Merck
All Photos(2)

Documents

ABS1668M

Sigma-Aldrich

Anti-CHMP3

serum, from rabbit

Synonym(s):

Charged multivesicular body protein 3, Chromatin-modifying protein 3, Neuroendocrine differentiation factor, Vacuolar protein sorting-associated protein 24, hVps24

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... CHMP3(51652)

General description

Charged multivesicular body protein 3 (UniProt: Q9Y3E7; also known as Chromatin-modifying protein 3, CHMP3, Neuroendocrine differentiation factor, Vacuolar protein sorting-associated protein 24, hVps24) is encoded by the CHMP3 (also known as CGI149, NEDF, VPS24, CGI-149) gene (Gene ID: 51652) in human. CHMP3 is a component of the endosomal sorting required for transport complex III (ESCRT-III), which is involved in multi-vesicular bodies (MVBs) formation and sorting of endosomal cargo proteins into MVBs. It is widely expressed and its expression is reported in heart, brain, placenta, lung, liver, skeletal muscle, kidney, and pancreas. The acidic C-terminus and the basic N-terminus of CHMP3 are thought to render the protein in a closed, soluble and inactive conformation through an auto-inhibitory intramolecular interaction. The open and active conformation, which enables membrane binding and oligomerization, is achieved by interaction with other cellular binding partners, probably including other ESCRT components. CHMP3 is shown to heteromerize with CHMP2A to form helical tubular structures that expose membrane-interacting sites on the outside. It can also interact with CHMP4A, however, the interaction requires the release of CHMP4A auto-inhibition.

Specificity

This rabbit polyclonal antibody detects Charged multivesicular body protein 3 in human cells.

Immunogen

A full-length human recombinant charged multivesicular body protein 3 (CHMP3).
Epitope: unknown

Application

Anti-CHMP3, Cat. No. ABS1668, is a rabbit polyclonal antibody that detects Charged multivesicular body protein 3 and has been tested for use in Immunocytochemistry and Western Blotting.
Research Category
Signaling
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected human recombinant charged multivesicular body protein 3 (CHMP3).

Western Blotting Analysis: A representative lot detected CHMP3 in Western Blotting applications (Morita, E., et. al. (2010). Proc Natl Acad Sci USA. 107(29):12889-94).

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected CHMP3 in HeLa cells.

Target description

~32 kDa observed; 25.07 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Rabbit polyclonal antiserum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service