Skip to Content
Merck
All Photos(1)

Key Documents

ABS1645

Sigma-Aldrich

Anti-Chromatin-Modifying Protein 6

from rabbit

Synonym(s):

Chromatin-modifying protein 6, Charged multivesicular body protein 6, Vacuolar protein sorting-associated protein 20, Vps20, hVps20

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, monkey

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CHMP6(79643)

General description

Charged multivesicular body protein 6 (UniProt: Q96FZ7; also known as Chromatin-modifying protein 6, Vacuolar protein sorting-associated protein 20, Vps20, hVps20, CHMP6) is encoded by the CHMP6 (also known as VPS20) gene (Gene ID: 79643) in human. CHMP6 is a human orthologue of yeast Vps (vacuolar protein sorting) 20, a component of ESCRT (endosomal sorting complex required for transport)-III. Various CHMP6 orthologues in organisms ranging from yeast to humans contain the N-myristoylation consensus sequence at each N-terminus. CHMP6 is an ubiquitously expressed protein that has an acidic C-terminus and the basic N-terminus that render the protein in a closed, soluble and inactive conformation through an auto-inhibitory intramolecular interaction. The open and active conformation, which enables membrane binding and oligomerization, is achieved by interaction with other cellular binding partners, probably including other ESCRT components. CHMP6 is probable core component of the endosomal sorting required for transport complex III (ESCRT-III), which is involved in multivesicular bodies (MVBs) formation and sorting of endosomal cargo proteins into MVBs. MVBs contain intraluminal vesicles that are generated by invagination and scission from the limiting membrane of the endosome and mostly are delivered to lysosomes enabling degradation of membrane proteins.

Specificity

This polyclonal antibody detects chromatin-modifying protein 6 (CHMP6) in human and monkey cells.

Immunogen

Epitope: unknown
GST-tagged full length recombinant human Chromatin-modifying protein 6.

Application

Anti-Chromatin-Modifying Protein 6, Cat. No. ABS1645, is a rabbit polyconal antibody that is used to detect Chromatin-Modifying Protein 6 and has been tested for use in Immunofluorescence and Western Blotting.
Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected Chromatin-Modifying Protein 6 in Cos-7 cells (Langelier, C., et. al. (2006). J Virol. 80(19):9465-80).

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Chromatin-Modifying Protein 6 in HEK293 cell lysate (Langelier, C., et. al. (2006). J Virol. 80(19):9465-80).
Research Category
Signaling

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: A 1:2,000 dilution of this antibody detected Chromatin-Modifying Protein 6 in 10 µg of HEK293 cell lysate.

Target description

~32 kDa observed; 23.48 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Format: Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service