Skip to Content
Merck
All Photos(2)

Documents

ABN468

Sigma-Aldrich

Anti-ASAH1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

ASAH1 Antibody, N-acylsphingosine amidohydrolase (non-lysosomal ceramidase) 1, acid ceramidase, hCG_28255

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ASAH1(427)

General description

N-acylsphingosine amidohydrolase (Acid ceramidase) 1, or ASAH1 is part of a family of hydrolyases, localized in the lysosome, that catalyze the degradation of ceramide into sphingosine (SPH) and free fatty acid. Ceraminde is a subgroup of sphingolipids and sphingolipids are important membrane lipids in all cells. Sphingolipids are being recognized today too as important second messengers in various signaling transduction pathways, principally via the release or use of free sphingosine. Because ceramide degradation is the only source of cellular SPH, these enzymes are not only important for limiting ceramide-mediated signaling but also for controlling the cellular functions of SPH and SPH-1 phosphate enzymes. Build up excess ceramides can have damaging effects on cells and indeed, ceramide addition is a potent inducer of apoptosis is most cell types. Defects in ASAH1 gene cause a condition called Farber lipogranulomatosis, also known as Farber disease (FD). This sphingolipid disease is characterized by subcutaneous lipid-loaded nodules, excruciating pain in the joints and extremities, marked accumulation of ceramide in lysosomes, and death by three years of age. Additionally, spinal muscular atrophy is also associated with defects in the ASAH1 protein which eventually results in death at an early age from respiratory failure. EMD-Millipore’s anti-ASAH1 rabbit polyclonal antibody has been tested in western blot on mouse heart tissue lysates, and by immunohistochemistry on human heart tissue, and by fluorescent immunocytochemistry on human heart cells in culture.

Immunogen

KLH-conjugated linear peptide corresponding to human ASAH1.

Application

Anti-ASAH1 Antibody is a highly specific rabbit polyclonal antibody, that targets ASAH1 & has been tested in western blotting, IHC & Immunofluorescence.
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected ASAH1 in human cardiac myocytes and kidney tissues.

Immunofluorescence Analysis: 20 µg/mL from a representative lot detected ASAH1 in human heart cells.
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blot in mouse heart tissue lysate.

Western Blot Analysis: 2 µg/mL of this antibody detected ASAH1 in 15 µg of mouse heart tissue lysate.

Target description

~52 kDa observed. Uncharacterized bands may appear in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse heart tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diego Iacono et al.
Neurobiology of disease, 174, 105862-105862 (2022-09-17)
Krabbe Disease (KD) is an autosomal recessive disorder that results from loss-of-function mutations in the GALC gene, which encodes lysosomal enzyme galactosylceramidase (GALC). Functional deficiency of GALC is toxic to myelin-producing cells, which leads to progressive demyelination in both the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service