Skip to Content
Merck
All Photos(3)

Documents

ABN1474

Sigma-Aldrich

Anti-RNF213

from rabbit, purified by affinity chromatography

Synonym(s):

E3 ubiquitin-protein ligase RNF213, EC:2.3.2.27, ALK lymphoma oligomerization partner on chromosome 17, Mysterin, RING finger protein 213, RING-type E3 ubiquitin transferase RNF213

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... RNF213(57674)

Related Categories

General description

E3 ubiquitin-protein ligase RNF213 (UniProt: Q63HN8; also known as EC:2.3.2.27, ALK lymphoma oligomerization partner on chromosome 17, Mysterin, RING finger protein 213, RING-type E3 ubiquitin transferase RNF213) is encoded by the RNF213 (also known as ALO17, C17orf27, KIAA1554, KIAA1618, MYSTR) gene (Gene ID: 57674) in human. RNF213 is a widely expressed, homooligomeric cytosolic protein that is involved in the non-canonical Wnt signaling pathway in vascular development. It acts by mediating ubiquitination and degradation of FLNA and NFATC2 downstream of RSPO3, which leads to the inhibition of the non-canonical Wnt signaling pathway and promotes vessel regression. Its activity is induced by pro-inflammatory cytokines. RNF213 contains a Zinc finger domain (aa 3997-4036), which is required for its ubiquitin-protein ligase activity. It also contains an AAA domain, which is associated with ATPase activity. Four isoforms of RNF213 have been described that are produced by alternative splicing. Mutations in RNF213 gene have been linked to Moyamoya disease 2 (MYMY2) disease, a progressive cerebral angiopathy that can lead to transient ischemic attacks and/or cerebral infarction, and rupture of the collateral vessels can cause intracranial hemorrhage.

Specificity

This rabbit polyclonal antibody detects E3 ubiquitin-protein ligase RNF213 in human cells. It targets an epitope within 15 amino acids from the C-terminal half.

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal half of human E3 ubiquitin-protein ligase RNF213. Immunogen sequence is conserved in isoforms 1, 2, and 4.

Application

Anti-RNF213, Cat. No. ABN1474, is a highly specific rabbit polyclonal antibody that targets E3 ubiquitin-protein ligase RNF213 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected RNF213 in human cerebral cortex and human kidney tissue sections.

Western Blotting Analysis: 2 µg/mL from a representative lot detected RNF213 in 10 µg of HEK293 cells transfected with 3XFLAG RNF213 WT.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in HUVEC lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected RNF213 in 10 µg of HUVEC lysate.

Target description

~590 kDa observed; 591.41 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service