Skip to Content
Merck
All Photos(2)

Key Documents

ABN1459

Sigma-Aldrich

Anti-Nkx6.1

from rabbit, purified by affinity chromatography

Synonym(s):

Homeobox protein Nkx-6.1, Homeobox protein NK-6 homolog A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... NKX6-1(4825)

General description

Homeobox protein Nkx-6.1 (UniProt: P78426; also known as Homeobox protein NK-6 homolog A) is encoded by the NKX6-1 (also known as NKX6A) gene (gene ID: 4825) in human. Nkx-6.1 is a transcription factor that binds to specific A/T-rich DNA sequences in the promoter regions of a number of genes. It is expressed in pancreatic beta cells and is involved in transcriptional regulation in islet beta cells. During pancreatic development both Pdx-1 and Nkx-6.1 are observed in rat pancreas. However, later Nkx-6.1 became more beta-cell-restricted, and Pdx-1 is found in other islet cell types and in the duodenal epithelium. Nkx-6.1 binds to the insulin promoter and is involved in regulation of the insulin gene. The C-terminal domain of Nkx-6.1 contributes to sequence-specific DNA-binding. Together with NKX2-2 and IRX3 it acts to restrict the generation of motor neurons to the appropriate region of the neural tube. Belongs to the class II proteins of neuronal progenitor factors, which are induced by Sonic hedgehog (Shh) signals. Although Shh signaling is required for Nkx-6.1 expression in the ventral neural tube and spinal meninges, a Shh-independent pathway appears to operate in regulating Nkx-6.1 expression in the foregut region. (Ref.: Cai, J., et al. (2000). Genesis. 27(1); 6-11; Oster, A., et al. (1998). J. Histochem. Cytochem. 46(6); 707-715).

Specificity

This rabbit polyclonal antibody detects Homeobox protein Nkx-6.1 in human cells. It targets an epitope within 61 amino acids from the N-terminal region.

Immunogen

GST/His-tagged recombinant fragment corresponding to 61 amino acids from the N-terminal region of human Homeobox protein Nkx-6.1.

Application

Anti-Nkx6.1, Cat. No. ABN1459, is a highly specific rabbit polyclonal antibody that targets Homeobox protein Nkx-6.1 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Nkx6.1 in human pancreas and mouse pancreas tissue sections.
Research Category
Neuroscience

Quality

Evaluated by Western Blotting in PANC-1 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Nkx6.1 in PANC-1 cell lysate.

Target description

~54 kDa observed; 37.85 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service