Skip to Content
Merck
All Photos(2)

Documents

ABN1367

Sigma-Aldrich

Anti-Pan DIDO Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Death-inducer obliterator 1, DATF-1, Death-associated transcription factor 1, DIDO1, DIDO2, DIDO3, DIO-1, hDido1, hDido2, hDido3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

nonhuman primates (based on 100% sequence homology), mouse (based on 100% sequence homology), Xenopus (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... DIDO1(11083)

General description

Death-inducer obliterator 1 (UniProt Q9BTC0; also known as DATF-1, Death-associated transcription factor 1, DIO-1, hDido1) is encoded by the DIDO1 (also known as C20orf158, DATF1, KIAA0333) gene (Gene ID 11083) in human. The DIDO1 gene encodes four alternatively spliced variants (DIDO1/isoform 2, DIDO2/isoform 1, DIDO3/isoform 4, isoform a/isoform 3) implicated to be involved in apoptosis and development. The longest and most widely expressed DIDO3 is assigned as the canonical isoform. DIDO3 is a nuclear protein that forms part of the spindle assembly checkpoint (SAC) and is necessary for correct chromosome segregation in somatic and germ cells. Dido3-knockout compromises differentiation of murine embryonic stem cells (mESCs) in vitro and of epiblast cells in vivo, resulting in early embryonic death at around day 8.5 of gestation. On the other hand, exogenous expression of Dido1 isoform in mESCs is reported to inhibit LIF withdrawal-induced differentiation, while Dido1 knockdown facilitate mESCs differentiation.

Specificity

This polyclonal antibody detects an N-terminal epitope present in all four spliced isoforms of human DIDO1 reported by UniProt (Q9BTC0).

Immunogen

KLH-conjugated linear peptide corresponding an N-terminal sequence of human DIDO1.

Application

Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected DIDO immunoreactivity in human kindey, human prostate cancer and human cerebellum tissue.
This Anti-Pan DIDO Antibody is validated for use in Western Blotting, Immunohistochemistry for the detection of DIDO.

Quality

Evaluated by Western Blotting in HepG2 cell lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected DIDO isoforms in 10 µg of HepG2 cell lysate.

Target description

~260/130/79/70/66 observed. 59.46 kDa (isoform 2; DIDO1), 129.2 kDa (isoform 1; DIDO2), 243.9 kDa (isoform 4; DIDO3), 61.35 kDa (isoform 3; isoform a) calculated. Uncharacterized band(s) may appear in some lysates.

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service