Skip to Content
Merck
All Photos(1)

Key Documents

ABE798

Sigma-Aldrich

Anti-KPNA6 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Importin subunit alpha-7, Karyopherin subunit alpha-6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KPNA6(23633)

General description

Importin subunit alpha-7, also known as Karyopherin subunit alpha-6, and encoded by the gene name KPNA6 or IPOA7, is a member of the Importin alpha family. KPNA6 is a cytosolic and heterodimeric protein complex consisting of alpha and beta subunits, is responsible for targeting proteins with nuclear localization signals to the nuclear pore complex (NPC) by an energy requiring, Randependent mechanism. The alpha subunit and imported substrate enter the nucleus and accumulate in the nucleoplasm, while the beta subunit accumulates at the NPC. KPNA6 belongs to a subfamily within the KPNA family that also includes IPOA5&6. Down-regulation of KPNA6 by RNAi expression in HeLa cells strongly inhibited cell proliferation, possibly due to blocking the nuclear import of specific factors essential for cell growth and proliferation. KPNA6 also interacts with the Rev protein of HIV-1 and promote its nuclear import

Specificity

This antibody is predicted to not cross-react with other Importin alpha family members.

Immunogen

KLH-conjugated synthetic peptide corresponding to the N-terminus of human KPNA6.

Application

Detect KPNA6 using this Anti-KPNA6 Antibody validated for use in western blotting & immunofluorescence.
Immunofluorescence Analysis: 20 μg/mL from a representative lot detected KPNA6 in HEK293 cells.

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1 μg/mL of this antibody detected KPNA6 in HEK293 cell lysate.

Target description

~63 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service