ABE484
Anti-LOXL2
from rabbit, purified by affinity chromatography
Synonym(s):
Lysyl oxidase homolog 2, EC: 1.4.3.13, Lysyl oxidase-like protein 2, Lysyl oxidase-related protein 2, Lysyl oxidase-related protein WS9-14
About This Item
Recommended Products
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
species reactivity (predicted by homology)
mouse (based on 100% sequence homology), rat (based on 100% sequence homology), bovine (based on 100% sequence homology)
packaging
antibody small pack of 25 μg
technique(s)
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
ambient
target post-translational modification
unmodified
Gene Information
human ... LOXL2(4017)
mouse ... Loxl2(94352)
General description
Specificity
Immunogen
Application
Epigenetics & Nuclear Function
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected LOXL2 in human prostate and human placenta tissues.
Immunoprecipitation Analysis: A representative lot detected LOXL2 in LoxL2 expressing vector (Courtesy Dr Sandra Peiro from the Institut Hospital del Mar d′Investigacions Mèdiques (IMIM) in Barcelona Spain ).
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected LOXL2 in human prostate and human placenta tissues.
Immunoprecipitation Analysis: A representative lot detected LOXL2 in LoxL2 expressing vector (Courtesy Dr Sandra Peiro from the Institut Hospital del Mar d′Investigacions Mèdiques (IMIM) in Barcelona Spain).
Quality
Western Blotting Analysis: A 1:500 dilution of this antibody detected LOXL2 in A431 cell lysate.
Target description
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service