Skip to Content
Merck
All Photos(2)

Key Documents

ABE1455

Sigma-Aldrich

Anti-Nurr1 (NR4A2)

from rabbit, purified by affinity chromatography

Synonym(s):

Nuclear receptor subfamily 4 group A member 2, Immediate-early response protein NOT, Orphan nuclear receptor NURR1, Transcriptionally-inducible nuclear receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... NR4A2(4929)

General description

Nuclear receptor subfamily 4 group A member 2 (UniProt: P43354; also known as Immediate-early response protein NOT, Orphan nuclear receptor NURR1, Transcriptionally-inducible nuclear receptor) and is encoded by the NR4A2 (also known as NOT, NURR1, TINUR) gene (Gene ID: 4929) in human. Nurr-1 belongs to the nuclear hormone receptor family that acts as a transcriptional regulator and is considered to be important for the differentiation and maintenance of meso-diencephalic dopaminergic (mdDA) neurons during development. It is also crucial for expression of a set of genes such as SLC6A3, SLC18A2, TH and DRD2 which are essential for development of mdDA neurons. It is expressed in a number of T cell, B cell and fibroblast cell lines, but its expression is higher in brain tissue. Two isoforms of Nurr-1 have been reported that are produced by alternative splicing. The ligand-binding domain (LBD) of Nurr-1 contains no cavity as a result of the tight packing of side chains from several bulky hydrophobic residues in the region normally occupied by ligands. It also lacks a classical binding site for co-activators. However, it has hydrophobic region on the surface of LBD, which is critical for its activity. (Ref.: Volakakis N et al (2006). J. Mol. Endocrinol 37(2); 317-326; Wang G et al. (2003). Nature 423(6939):555-560).

Specificity

This rabbit polyclonal antibody detects NR4A2 (Nurr1) in human cells. It targets an epitope within 16 amino acids from the N-terminal region.

Immunogen

Epitope: N-terminus
KLH-conjugated liner peptide corresponding to 16 amino acids from the N-terminal region of human Nuclear receptor subfamily 4 group A member 2 (Nurr1).

Application

Anti-Nurr1 (NR4A2), Cat. No. ABE1455, is a highly specific rabbit polyclonal antibody that targets Nuclear receptor subfamily 4 group A member 2 and has been tested for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Nurr1 (NR4A2) in HeLa cells.
Research Category
Epigenetics & Nuclear Function

Quality

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected Nurr1 (NR4A2) in THP-1 cell lysate.

Target description

~67 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carl G Gottschalk et al.
Molecular neurobiology, 56(11), 7872-7887 (2019-05-28)
Nuclear receptor related 1 protein (Nurr1) is an important transcription factor required for differentiation and maintenance of midbrain dopaminergic (DA) neurons. Since decrease in Nurr1 function either due to diminished expression or rare mutation is associated with Parkinson's disease (PD)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service