Skip to Content
Merck
All Photos(2)

Key Documents

ABC521

Sigma-Aldrich

Anti-Rab21 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Ras-related protein Rab-21

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human, rat

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... RAB21(23011)

General description

RAB21 is a member of the small GTPase superfamily that is critically important in the recycling of integrin protein receptor complexes and their internalization. Thus, RAB21 is important in cell adhesion and migration of cells, as well as the proper completing of cytokinesis which requires particular care with adherent cells to coordinate the release and recycling of integrin and their release from the ECM matrix during cytokinesis. RAB21 is primarily a cytoplasmic or cytoplasmic organelle localized protein and its precise location will vary with cell cycle and cell division phases. RAB21 is widely expressed particularly in cells in the gut and capillaries where cell division and cell morphological changes are common.

Immunogen

GST-tagged recombinant protein corresponding to Mouse Rab21.

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-Rab21 Antibody is validated for use in Western Blotting for the detection of Rab21.
Western Blot Analysis: A 1:1000 dilution of a representative lot of this antibody detected Rab21 in A431, C2C12, HEK293, HeLa, HepG2, Jurkat, L6, Mouse brain, NIH/3T3, PC12, PC3, and RAW264.7 cell lysates. This antibody did not detect Rab21 in Human placenta or Huvec cell lysates.

Western Blot Analysis: A representative lot of this antibody detected Rab21 in melanocyte membrane compartment lysate (Ohbayashi et al., (2012) Mol Biol Cell. 23(4):669-678).

Quality

Evaluated by Western Blotting in PC12 cell lysate.

Western Blotting Analysis: A 1:1000 dilution of this antibody detected Rab21 in 10 µg of PC12 cell lysate.

Target description

~24 kDa observed.
Uncharacterized bands may be obeserved in some lysates.

Physical form

Antigen Affinity Purified
Purified rabbit Polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service