Skip to Content
Merck
All Photos(1)

Key Documents

AB9485

Sigma-Aldrich

Anti-Somatostatin Receptor Type 1 Antibody

Chemicon®, from rabbit

Synonym(s):

SSTR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SSTR1(6751)

Specificity

Recognizes Somatostatin Receptor Type 1 [SSTR1].

Immunogen

Synthetic peptide from the N-terminal extracellular domain of human Somatostatin Receptor Type 1.

Application

Anti-Somatostatin Receptor Type 1 Antibody is an antibody against Somatostatin Receptor Type 1 for use in IH(P).
Immunohistochemistry on paraffin embedded tissue sections.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Hormones & Receptors

Physical form

Affinity purified immunoglobulin. Liquid in PBS, pH 7.7 with 0.01% sodium azide.

Storage and Stability

Maintain at -70°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weike Liu et al.
Evidence-based complementary and alternative medicine : eCAM, 2022, 5910874-5910874 (2022-08-23)
Exosomes derived from human bone marrow mesenchymal stem cells (BMSCs) play potential protective roles in asthma. However, the underlying mechanisms remain not fully elucidated. Herein, exosomes were isolated from BMSCs, and the morphology, particle size, and exosome marker proteins were
Gaoyang Chen et al.
International journal of biological sciences, 17(15), 4409-4425 (2021-11-23)
Bone remodeling is a dynamic process between bone formation mediated by osteoblasts and bone resorption mediated by osteoclasts. Disrupted bone remodeling is a key factor in postmenopausal osteoporosis, a metabolic disorder characterized by deteriorated bone microarchitecture and increased risk of
Kathleen E Morrison et al.
Endocrinology, 157(5), 2002-2014 (2016-03-05)
The peripubertal period of development is a sensitive window, during which adverse experiences can increase the risk for presentation of cognitive and affective dysfunction throughout the lifespan, especially in women. However, such experiences in the context of a supportive social
Monika Maleszewska et al.
Angiogenesis, 19(1), 9-24 (2015-09-30)
High uniform fluid shear stress (FSS) is atheroprotective and preserves the endothelial phenotype and function through activation of downstream mediators such as MAPK7 (Erk5). Endothelial cells respond to FSS thanks to mechanotransduction. However, how the resulting signaling is integrated and
Shijie Zhang et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 47(2), 667-679 (2018-05-26)
In this study, we aimed to use bioinformatics tools to identify the specific miRNAs and mRNAs involved in osteogenic differentiation and to further explore the way in which miRNA regulates osteogenic differentiation. The microarray GSE80614, which includes data from 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service