Skip to Content
Merck
All Photos(2)

Documents

AB4330

Sigma-Aldrich

Anti-DDX4 (Vasa) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

DEAD (Asp-Glu-Ala-Asp) box polypeptide 4, probable ATP-dependent RNA helicase DDX4, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 4, Vasa homolog, DEAD box protein 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DDX4(54514)

General description

DDX4 (Vasa or MVH) is a member of the DEAD family of proteins and is a probable ATP-dependent RNA helicase. DDX4 (Vasa) proteins are mostly localized in testicular germ cells in the cytoplasm of spermatocytes. DDX4 (Vasa) is a high-copy extragenic suppressor gene. DDX4 (Vasa) is shown to be significantly decreased in spermatozoa of men that are infertile and may have an important role in male fertility.

Specificity

This antibody recognizes DDX4 (Vasa) at the C-terminus.

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of mouse DDX4 (Vasa).

Application

Anti-DDX4 (Vasa) Antibody detects level of DDX4 (Vasa) & has been published & validated for use in WB, IH(P).
Immunohistochemistry Analysis: A 1:100-250 dilution from a previous lot detected DDX4 (Vasa) in normal human testis tissue.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

RNA Metabolism & Binding Proteins

Quality

Evaluated by Western Blot in mouse testis tissue lysate.

Western Blot Analysis: A 1:500 dilution of this antibody detected DDX4 (Vasa) in 10 µg of mouse testis tissue lysate. Western Blot performed without Tween.

Target description

~76 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse testis tissue lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Irma Virant-Klun et al.
Cells, 8(7) (2019-07-25)
A population of small stem cells with diameters of up to 5 μm resembling very small embryonic-like stem cells (VSELs) were sorted from human embryonic stem cell (hESC) cultures using magnetic-activated cell sorting (MACS) based on the expression of a
Tzu-Bou Hsieh et al.
Journal of ovarian research, 17(1), 37-37 (2024-02-10)
Premature ovarian insufficiency (POI) is a condition defined as women developing menopause before 40 years old. These patients display low ovarian reserve at young age and difficulties to conceive even with assisted reproductive technology. The pathogenesis of ovarian insufficiency is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service