Skip to Content
Merck
All Photos(1)

Documents

AB3349

Sigma-Aldrich

Anti-PRMT5 Antibody, CT

Chemicon®, from rabbit

Synonym(s):

Anti-HRMT1L5, Anti-HSL7, Anti-IBP72, Anti-JBP1, Anti-SKB1, Anti-SKB1Hs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PRMT5(10419)

Specificity

Reacts with PRMT5.

Immunogen

Epitope: C-terminus
Synthetic peptide from C-terminus of human PRMT5.

Application

Anti-PRMT5 Antibody, C-terminus is a high quality Rabbit Polyclonal Antibody for the detection of PRMT5 & has been validated in ELISA & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
Western Blot: 1:100-1:500. Reacts with a band of approximately 70 kDa on whole HL60 cell lysate. Additional bands may be present depending on sample used.

ELISA (direct): 1:1,000

Optimal working dilutions must be determined by the end user.

Linkage

Replaces: MABE551

Physical form

Format: Purified
Purified immunoglobulin in PBS containing 0.09% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
POSITIVE CONTROL: HL60 cell lysates.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service