Skip to Content
Merck
All Photos(2)

Documents

AB3256-I

Sigma-Aldrich

Anti-Luciferase

from rabbit, purified by affinity chromatography

Synonym(s):

Luciferin 4-monooxygenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity (predicted by homology)

all

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

Photinus pyralis ... Loc116160065(116160065)

General description

Luciferin 4-monooxygenase (UniProt: P08659; also known as EC: 1.13.12.7, Luciferase) is a peroxisomal enzyme that belongs to the ATP-dependent AMP-binding enzyme family. It catalyzes the production of oxyluciferin from luciferin in an ATP and magnesium dependent reaction and releases a photon of light when oxyluciferin returns to the ground state. This reaction proceeds in two steps where the first step involves adenylation of luciferin and in the second step luciferase acts as an oxygenase on adenyl-luciferin to produce oxyluciferin and carbon dioxide, the decay of oxyluciferin producing a photon of light of 562 nm. Luciferase from the firefly is commonly used in reporter assay where the total light emission is proportional to the luciferase activity of the sample.

Specificity

This rabbit polyclonal antibody detects firefly luciferase.

Immunogen

Epitope: unknown
GST-tagged full length recombinant firefly luciferase.

Application

Anti-Luciferase, Cat. No. AB3256-I, is a rabbit polyclonal antibody that detects firefly luciferase and has been tested for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:500 dilution from a representative lot detected Luciferase in HeLa cells transfected with Luciferase expression plasmid.
Research Category
Secondary & Control Antibodies

Quality

Evaluated by Western Blotting in lysate from HeLa cells transfected with Luciferase expression plasmid.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Luciferase in HeLa cells transfected with luciferase expression plasmid.

Target description

~58 kDa observed; 60.75 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service