Skip to Content
Merck
All Photos(1)

Key Documents

AB3121

Sigma-Aldrich

Anti-Angiopoietin-2 Antibody, NT

Chemicon®, from rabbit

Synonym(s):

Ang-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

mouse ... Angpt2(11601)

Specificity

Recognizes mouse Angiopoietin-2 (Ang-2). Mouse/human Ang-2 is a 496 AA secreted angiogenic protein (Suri et al. 1996). There is ~85% homology between mouse and human Ang-2. It is only 60% homologous with Ang-1 (Davis et al. 1996). Ang-2 may act as an antagonist of Ang-1 and Tie-2 (Suri et al. 1996). It is prominently expressed in tissues that are in involved in vascular remodeling (ovary, placenta, and uterus).

The immunogenic peptide sequence is 71% (15/21 aa) homologous with human Ang-2 and has no substantial homology with Ang-1. Antibody crossreactivity in other species is not established. The immunogen peptide is 85% conserved in porcine and 55% in chicken.

Immunogen

20 amino acid peptide sequence near the amino-terminus of mouse Ang-2.
Epitope: N-terminus

Application

Detect Angiopoietin-2 using this Anti-Angiopoietin-2 Antibody, N-terminus validated for use in ELISA & WB.
Research Category
Cell Structure
Research Sub Category
ECM Proteins
Western blot: 1-10 μg/mL using ECL. The predicted size of Ang-2 is ~55 kDa. Like Ang-1, its size may higher due to glycosylation.

ELISA: 1:10,000-1:100,000 using 50-100 ng of control peptide per well.

Optimal working dilutions must be determined by end user.

Physical form

Affinity purified immunoglobulin presented as liquid in PBS containing 0.1% BSA and 0.05% azide, pH 7.5

Storage and Stability

Maintain frozen at -20°C in undiluted aliquots for up to 12 months from date of receipt. May be stored at 2-8°C for short term use. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Durham-Lee et al.
Neuroscience, 202, 454-464 (2011-10-25)
Angiopoietin-1 (Ang-1) and angiopoietin-2 (Ang-2) have opposing effects on blood vessels, with Ang-2 being mainly induced during the endothelial barrier breakdown. It is known that spinal cord injury (SCI) induces lasting decreases in Ang-1 levels, underlying endothelial barrier disruption, but
Sunil K Chauhan et al.
Arteriosclerosis, thrombosis, and vascular biology, 35(7), 1606-1615 (2015-05-23)
In angiogenesis, circulating mononuclear cells are recruited to vascular lesions; however, the underlying mechanisms are poorly understood. Here, we characterize the functional role of protein tyrosine kinase 7 (PTK7)-expressing CD11b(+) mononuclear cells in vitro and in vivo using a mouse

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service