Skip to Content
Merck
All Photos(1)

Documents

AAWP09050

Millipore

MF-Millipore Membrane Filter, 0.8 µm pore size

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602

material

plain white mixed cellulose esters (MCE) filter

feature

hydrophilic

manufacturer/tradename

MF-Millipore

parameter

max. temp. at 75 °C
16 L/min-cm2 air flow rate
190 mL/min-cm2 water flow rate

filter diam. × thickness

90 mm × 180 μm

gravimetric extractables

4%

refractive index

n/D 1.51

pore size

0.8 μm pore size

bubble point

≥1.10 bar, air with water

General description

AAWP
Biologically inert mixtures of cellulose acetate and cellulose nitrate have made MF-Millipore membrane filters one of the most widely used membranes in analytical and research applications.

MF-Millipore filters without Triton surfactant contain minimum amounts of wetting agent and have a lower water extractable content than standard MF-Millipore filters.

Features & Benefits:
- Versatile filter for biological and environmental monitoring applications
- Available in a range of pore sizes, colored black or white, with or without a gridded surface
- Compatible with ethylene oxide, gamma irradiation, and autoclave sterilization methods
Screen filter

Application

  • Air monitoring
  • Particle monitoring
  • Particle removal
  • Bioassays
  • Microplastics

Legal Information

MF-Millipore is a trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service