Skip to Content
Merck
All Photos(1)

Key Documents

964205

Supelco

Reducing valve, 0.2 bar with 2 tube connections 6 x 8 mm and integrated excess pressure safety device 0.5 bar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NB.51
Pricing and availability is not currently available.

shipped in

ambient

storage temp.

no temp limit

General description

The pressure reducer with included overpressure relief is one of the safest combined solutions for reducing inert gas house lines to a working pressure of 0.2 bar. The integrated overpressure relieve avoids pressures higher than 0.5 bar inside the barrel according the Pressure Equipment Directive. The pressure reducer is equipped with a ball valve and with 2 tube connectors 8x6 mm.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service