Skip to Content
Merck
All Photos(1)

Key Documents

9031

Millipore

Minicon® - B15 Static Concentrator

8 cells/unit, 15 kDa MWCO, 5 mL sample volume

Synonym(s):

Minicon B15, Minicon Static Concentrator, Static Concentrator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
34360190

manufacturer/tradename

Minicon®

parameter

5 mL sample volume

pore size

15 kDa NMWCO

shipped in

ambient

General description

Overview

  • Concentrate urine and cerebrospinal fluid to intensify proteins that indicate abnormal or pathological states prior to analysis by electrophoresis or immunoelectrophoresis.
  • Static concentrator, requiring no accessories
  • Absorbent pulls solvent and salts through ultrafilter, concentrating sample
Filter Type: Ultrafiltration
Use the Minicon static concentrators to concentrate urine and cerebrospinal fluid to intensify proteins that indicate abnormal or pathological states prior to analysis by electrophoresis or immunoelectrophoresis. The B15 unit has 8 cells and can hold up to 5 mL of sample. The CS15 has 10 cells and is suited for volumes up to 2.5 mL.

Applications:
Concentration of Urine and Cerebrospinal Fluid for Enrichment of Protein Analytes.

Features and Benefits

Minimum Final Concentrate Volume: 50 µL

Static concentrator, requiring no accessories

Absorbent pulls solvent and salts through ultrafilter, concentrating sample

Concentration factor up to 100X

Low hold-up volume

Physical form

Color Code: Clear

Legal Information

MINICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service