Skip to Content
Merck
All Photos(1)

Key Documents

869031-M

Sigma-Aldrich

Chorionic Gonadotropin, Human Urine, Iodination Grade

Synonym(s):

Chorionic Gonadotropin, Human Urine, Iodination Grade

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$738.00

R$738.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
100 μG
R$738.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77
biological source:
human
form:
solid

R$738.00


Estimated to ship onJune 01, 2025


Request a Bulk Order

biological source

human

Quality Level

description

Merck USA index - 14, 2216

sterility

non-sterile

Assay

>99% (weight)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: soluble

storage temp.

−20°C

General description

Native, human chorionic gonadotropin from urine. Glycoprotein hormone synthesized by chorionic tissue of the placenta and found in urine during pregnancy. Also present in body fluids of patients with trophoblastic disease and ovarian tumors. Immunopotency: ≥10,000 IU/mg (WHO 3rd IS 75/537).

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized solid containing 25 mM NH₄HCO₃.

Preparation Note

Prepared from urine that has been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

Customers in Europe may require an import licence to purchase this product.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service