Skip to Content
Merck
All Photos(1)

Documents

8.55104

Sigma-Aldrich

2-(4-Bromomethylphenoxy)ethyl polystyrene HL

Novabiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111023
NACRES:
NA.22

Quality Level

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

General description

This versatile support is an excellent tool for the solid phase immobilization of carboxylic acids, amines and alcohols. Acids are attached by treating the resin with the appropriate DIPEA salt in DMF in the presence of CsI [1,2]. Reaction with aryl and alkyl amines provides resin-bound secondary amines. These can be readily acylated to give sulfonamides or anilides that can be cleaved with 95% TFA. Since this resin does not contain a benzylic ether linkage, it gives cleaner products than similar supports derived from Wang resin, free from impurities arising from breakdown of the linker. For examples using a similar resin, please see [1,2,3,4].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] J. W. Corbett, et al. (1997) Bioorg. Med. Chem. Lett., 7,1371.
[2] G. A. Morales, et al. (1998) J. Org. Chem., 63,1172.
[3] K. Ngu & D. V. Patel (1997) Tetrahedron Lett., 38, 973.
[4] B. Raju & T. P. Kogan (1997) Tetrahedron Lett., 38, 4965.

Linkage

Replaces: 01-64-0400

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Swelling Volume (in DMF): lot specific result
Loading (determined by elemntal analysis of bromine): 0.80 - 1.30 mmol/g
The polymer matrix is copoly (styrene - 1 % DVB), 100 - 200 mesh

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service