Skip to Content
Merck
All Photos(1)

Documents

8.55058

Sigma-Aldrich

Universal PEG NovaTag®_resin

Novabiochem®

Synonym(s):

N-Fmoc-NÆ-Mmt-PEG-diamine-MPB-AM resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51171641

product line

NovaSyn® TG
Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

15-25°C

General description

This resin has recently been used to prepare Shc and Src homology domain binding peptoid-peptide hybrids [1] and dimeric SMAC PEG-linked peptides [2].

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Literature references

[1] W. J. Choi (2009) J. Med. Chem., 52, 1612.
[2] Z. Gao, et al. (2007) J. Biol. Chem., 282, 30718.

Linkage

Replaces: 04-12-3911

Analysis Note

Color (visual): white to light yellow to beige
Appearance of substance (visual): beads
Loading (Photometric determination of the Fmoc-chromophore liberated upon treatment with DBU/DMF): 0.20 - 0.50 mmol/g
Swelling Volume (in DMF): lot specific result

Legal Information

NOVASYN is a registered trademark of Merck KGaA, Darmstadt, Germany
NOVATAG is a registered trademark of Merck KGaA, Darmstadt, Germany
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Chromogenic and fluorogenic derivatives are invaluable tools for biochemistry, having numerous applications in enzymology, protein chemistry, immunology and histochemistry.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service